missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dcp1a Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-04039CL1
This item is not returnable.
View return policy
Description
Dcp1a Polyclonal antibody specifically detects Dcp1a in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Dcp1a | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55802 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| DCP1 decapping enzyme homolog A (S. cerevisiae), decapping enzyme hDcp1a, EC 3.-, FLJ21691, mRNA-decapping enzyme 1A, Nbla00360, putative protein product of Nbla00360, Smad4-interacting transcriptional co-activator, SMAD4IP1, SMIFHSA275986, Transcription factor SMIF | |
| The immunogen this antibody was made to, maps to a region between residue 475 and 525 of human Dcp1a Antibody using the numbering given in entry NP_060873.4 (GeneID 55802). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction