missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCP1B Monoclonal Antibody (OTI1G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809931
This item is not returnable.
View return policy
Description
DCP1B is a core component of the mRNA decapping complex, a key factor in the regulation of mRNA decay.Specifications
| DCP1B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IZD4 | |
| DCP1B | |
| Human recombit protein fragment corresponding to amino acids 167-473 of human DCP1B produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1G8 | |
| Unconjugated | |
| DCP1B | |
| DCP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 196513 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction