missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCP1B Monoclonal Antibody (OTI1G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809931S
This item is not returnable.
View return policy
Description
DCP1B Monoclonal antibody specifically detects DCP1B in Human samples. It is validated for Western BlotSpecifications
| DCP1B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IZD4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 196513 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1G8 | |
| Unconjugated | |
| DCP1B | |
| B930050E02Rik; DCP1; DCP1 decapping enzyme homolog b; DCP1 decapping enzyme homolog B (S. cerevisiae); Dcp1b; decapping enzyme hDcp1b; decapping mRNA 1B; hDcp1b; mRNA-decapping enzyme 1B; RGD1562214 | |
| Human recombinant protein fragment corresponding to amino acids 167-473 of human DCP1B produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction