missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DCP1B Polyclonal Antibody
GREENER_CHOICE

Product Code. 16351345
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16351345 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16351345 Supplier Invitrogen™ Supplier No. PA584392

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VRSLSYEEPR RHSPPIEKQL CPAIQKLMVR SADLHPLSEL PENRPCENGS THSAGEFFTG PVQPGSPHNI GTSRGVQNAS RTQNL Highest antigen sequence indentity to the following orthologs: Mouse - 52%, Rat - 49%.

DCP1B is a core component of the mRNA decapping complex, a key factor in the regulation of mRNA decay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCP1B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCP1B
Gene Accession No. Q8IZD4
Gene Alias B930050E02Rik; DCP1; DCP1 decapping enzyme homolog b; DCP1 decapping enzyme homolog B (S. cerevisiae); Dcp1b; decapping enzyme hDcp1b; decapping mRNA 1B; hDcp1b; mRNA-decapping enzyme 1B; RGD1562214
Gene Symbols DCP1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DCP1B. Recombinant protein control fragment (Product #RP-106889).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196513
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.