missing translation for 'onlineSavingsMsg'
Learn More

DCPS Monoclonal Antibody (OTI4H8), TrueMAB™, OriGene

Product Code. p-200013107
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17574342 30 μL 30µL
17554342 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17574342 Supplier Origene Technologies Supplier No. TA505626S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DCPS Monoclonal antibody specifically detects DCPS in Canine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen DCPS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene GLB1
Gene Accession No. P16278, P23780, Q9TRY9
Gene Alias 5'-(N(7)-methyl 5'-triphosphoguanosine)-[mRNA] diphosphatase; ARS; Dcps; DCS1; DCS-1; decapping enzyme, scavenger; decapping scavenger enzyme; heat shock-like protein 1; HINT5; HINT-5; Hint-related 7meGMP-directed hydrolase; Histidine triad nucleotide-binding protein 5; histidine triad protein member 5; homolog of C. elegans 7meGMP-directed hydrolase dcs-1; HSL1; HSPC015; m7GpppX diphosphatase; mRNA decapping enzyme; scavenger mRNA-decapping enzyme DcpS
Host Species Mouse
Immunogen Full length human recombinant protein of human GLB1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12091, 2720, 316033, 403873
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.