Learn More
DCPS Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28465-1-AP-20UL
 View more versions of this product
Product Code. 18028755
  • € 145.00 / 20µL
Description
The DCPS antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DCPS. This antibody recognizes human, mouse antigen. The DCPS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
DCPS
280 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, ELISA
DCPS, DCS 1, DCS1, decapping enzyme, scavenger, HINT 5, HINT5, HSL1, HSPC015
20 μL
28960
Store at -20°C. Stable for one year after shipment.