missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DcR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585401
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Bovine (84%). Positive Control: 293T, A431, Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.
Specifications
| DcR3 | |
| Polyclonal | |
| Unconjugated | |
| TNFRSF6B | |
| DcR3; Decoy receptor 3; decoy receptor 3 variant 1; decoy receptor 3 variant 2; decoy receptor for Fas ligand; DJ583P15.1.1; hCG_2044836; M68; M68E; OTTHUMP00000031583; TNF receptor superfamily member 6b; TNFRSF6B; TR6; tumor necrosis factor receptor superfamily member 6B; tumor necrosis factor receptor superfamily member 6b, decoy; tumor necrosis factor receptor superfamily, member 6b, decoy; UNQ186/PRO212 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8771 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.87 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O95407 | |
| TNFRSF6B | |
| Recombinant protein encompassing a sequence within the center region of human DcR3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction