missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DCST1 Monoclonal Antibody (2C12)
GREENER_CHOICE

Product Code. 15543182
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15543182 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15543182 Supplier Invitrogen™ Supplier No. MA523086

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: MESNNMPCLP QPVGLDARAY WRAAVPIGLL VCLCLLQAFG YRLRRVIAAF YFPKREKKRI LFLYNDLLKK RAAFTKLRRA AILRRERQQK APRHPLADI.

This gene encodes a protein with a domain similar to one found in dendritic cells (PMID:11169400) which play a key role in antigen processing and display for immune responses. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCST1
Applications ELISA
Classification Monoclonal
Clone 2C12
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene DCST1
Gene Accession No. Q5T197
Gene Alias A330106H01Rik; Dcst1; DC-STAMP domain containing 1; DC-STAMP domain-containing protein 1; E3 ubiquitin-protein ligase DCST1; MLL5; RGD1307756; RING-type E3 ubiquitin transferase
Gene Symbols DCST1
Host Species Mouse
Immunogen DCST1 (NP_689707, 532 a.a. ~ 630 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149095
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.