Learn More
DCXR Antibody (OTI7D11), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71906F
Product Code. 18171993
  • € 751.00 / 0.10mL
Description
DCXR Monoclonal specifically detects DCXR in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DCXR
Monoclonal
FITC
PBS with 0.05% Sodium Azide
DCXR
Full length recombinant protein of human DCXR (NP_057370) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG1
Immunohistochemistry
OTI7D11
Immunohistochemistry
Carbonyl reductase II, DCR, dicarbonyl/L-xylulose reductaseP34H, EC 1.1.1, EC 1.1.1.10, HCR2, HCRII, human carbonyl reductase 2, KIDCR, Kidney dicarbonyl reductase, L-xylulose reductase, SDR20C1, short chain dehydrogenase/reductase family 20C, member 1, Sperm surface protein P34H, XR
Mouse
Affinity Purified
Apoptosis, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Lipid and Metabolism
51181.0
Store at 4°C in the dark.