missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDA1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16686944
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16686944 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16686944 Supplier Invitrogen™ Supplier No. PA5100465

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DDA1.

Functions as a component of numerous distinct DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complexes which mediate the ubiquitination and subsequent proteasomal degradation of target proteins (PubMed:17452440, PubMed:28437394, PubMed:28302793, PubMed:31686031, PubMed:31819272). In the DCX complexes, acts as a scaffolding subunit required to stabilize the complex (PubMed:31686031, PubMed:31819272). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDA1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DDA1
Gene Accession No. Q9BW61, Q9D9Z5
Gene Alias 1500034J01Rik; 1700095J19Rik; C19orf58; cross-immune reaction antigen PCIA1; Dda1; DET1 and DDB1 associated 1; DET1- and DDB1-associated protein 1; PCIA1; PCIA-1; Placenta cross-immune reaction antigen 1; R74921
Gene Symbols DDA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DDA1(Accession Q9BW61), corresponding to amino acid residues K65-T102.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66498, 688813, 79016
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.