missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDAH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550610
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total DDAH1 protein.
This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity.
Specifications
| DDAH1 | |
| Polyclonal | |
| Unconjugated | |
| DDAH1 | |
| 2410006N07Rik; 2510015N06Rik; AI987801; AW050362; Ddah; Ddah1; DDAH-1; DDAHI; dimethylargininase-1; dimethylarginine dimethylaminohydrolase 1; epididymis secretory protein Li 16; FLJ21264; FLJ25539; HEL-S-16; N(G),N(G)-dimethylarginine dimethylaminohydrolase 1; NG, NG-dimethylarginine dimethylaminohydrolase; NG,NG dimethylarginine dimethylaminohydrolase; RP4-621F18.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23576, 64157, 69219 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 3.1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O08557, O94760, Q9CWS0 | |
| DDAH1 | |
| Synthetic peptide corresponding to a region derived from internal residues of human dimethylarginine dimethylaminohydrolase 1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction