missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: SKRTNQRGAE ILADTFKDYA VSTVPVADGL HLKSFCSMAG PNLIAIGSSE SAQKALKIMQ QMSDHRYDKL TVPDDIAANC IYLNIPNKGH VLLHRTPEEY PESAKVYEKL KDHMLIPVSM SELEKVDGLL T Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity.
Specifications
Specifications
| Antigen | DDAH1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.20 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DDAH1 |
| Gene Accession No. | O08557, O94760, Q9CWS0 |
| Gene Alias | 2410006N07Rik; 2510015N06Rik; AI987801; AW050362; Ddah; Ddah1; DDAH-1; DDAHI; dimethylargininase-1; dimethylarginine dimethylaminohydrolase 1; epididymis secretory protein Li 16; FLJ21264; FLJ25539; HEL-S-16; N(G),N(G)-dimethylarginine dimethylaminohydrolase 1; NG, NG-dimethylarginine dimethylaminohydrolase; NG,NG dimethylarginine dimethylaminohydrolase; RP4-621F18.1 |
| Gene Symbols | DDAH1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
