missing translation for 'onlineSavingsMsg'
Learn More

DDB1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-98785C

Product Code. 18072588

  • € 415.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DDB1 Polyclonal specifically detects DDB1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DDB1
Polyclonal
50 mM Sodium Borate
Rabbit
Antigen Affinity-purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 650
damage-specific DNA binding protein 1 (127kD), damage-specific DNA binding protein 1, 127kDa, Damage-specific DNA-binding protein 1, DDB p127 subunit, DDBa, DNA damage-binding protein 1, DNA damage-binding protein a, HBV X-associated protein 1, UV-damaged DNA-binding factor, UV-damaged DNA-binding protein 1, UV-DDB 1, UV-DDB1, XAP1, XAP-1, Xeroderma pigmentosum group E-complementing protein, XPCe, XPE, XPE-BF, XPE-binding factor
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human DDB1.
0.1 mL
DNA Repair, Nucleotide Excision Repair
1642
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.