missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDB1 Mouse, anti-Human, CoraLite Plus 488, Clone: 4F6C6, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | DDB1 |
|---|---|
| Clone | 4F6C6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18050316
|
Proteintech
CL488-66010-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18020496
|
Proteintech
CL488-66010-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DDB1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DDB1. This antibody recognizes human, mouse, rat antigen. The DDB1 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| DDB1 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 1642 | |
| IgG2b | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 4F6C6 | |
| Western Blot | |
| Liquid | |
| Human, Mouse, Rat | |
| DDB p127 subunit, DDB1, DDBA, DNA damage binding protein 1, DNA damage binding protein a, HBV X associated protein 1, UV damaged DNA binding factor, UV DDB 1, UV DDB1, XAP 1, XAP1, XPCE, XPE, XPE BF, XPE binding factor | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title