missing translation for 'onlineSavingsMsg'
Learn More

DDB2 Monoclonal Antibody (OTI2E12), TrueMAB™, OriGene

Product Code. 17514134
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17514134 30 μL 30µL
17594124 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17514134 Supplier Origene Technologies Supplier No. TA810712S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DDB2 Monoclonal antibody specifically detects DDB2 in Human samples. It is validated for Western Blot

Specifications

Antigen DDB2
Applications Western Blot
Classification Monoclonal
Clone OTI2E12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FOXO1
Gene Accession No. Q12778
Gene Alias 2610043A19Rik; damage specific DNA binding protein 2; damage-specific DNA binding protein 2, 48kDa; damage-specific DNA-binding protein 2; DDB p48 subunit; DDB2; DDBb; DNA damage-binding protein 2; UV-damaged DNA-binding protein 2; UV-DDB 2; UV-DDB2; xeroderma pigmentosum group E protein; XPE
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 356-655 of human FOXO1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2308
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.