Learn More
DDHD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-13903
 View more versions of this product
Product Code. 18701144
  • ONLINE EXCLUSIVE PRICE
    € 589.00
    € 556.50 / 0.10mL
    Save € 32.50
    5% Off
Description
DDHD1 Polyclonal specifically detects DDHD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunohistochemistry
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Rabbit
IgG
Polyclonal
DDHD1
This antibody was developed against a recombinant protein corresponding to the amino acids: NTAMMREAARKIEERHFSNHATHVEFLPVEWRSKLTLDGDTVDSITPDKVRGLRDMLNSSAMDIMYYTSPLYRDELVKGLQQELNRLYSLFCSRNPD