Learn More
Invitrogen™ DDI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589849
Description
Immunogen sequence: SGIAVPGTSS SRPQHPGQQQ QRTPAAQRSQ GLASGEKVAG LQGLGSPALI RSMLLSNPHD LSLLKERNPP LAEALLSGSL ETFSQVLMEQ QREKALREQE RLRLYTADPL DREAQAKIEE EIRQQNIEEN MNIAIEEAPE SFGQVTMLYI NCKVNGHPLK AFVDSGAQMT IMSQACAERC NIMRLVDRRW AGVAKGVGTQ RIIGRVHLAQ IQIEGDFLQC SFSILEDQPM DMLLGLDMLR RHQCSIDLKK NVLVIGTTGT QTYFLPEGEL PLCSRMVSGQ DESSDKEITH SVMDSGRKEH; Positive Samples: Mouse testis.
Ddi1 (DNA-damage inducible 1)appears to release substrate from an ubiquitination complex, making the substrate available for deubiquitination to facilitate proteasomal degradation. Ddi1 could be a potential selective target for lung cancer treatment. Ddi1 runs as a doublet of 54/56 kDa on acrylamide gels besides the calculated MW 44 kDa.
Specifications
| DDI1 | |
| Polyclonal | |
| Unconjugated | |
| DDI1 | |
| 1700011N24Rik; DDI1; DDI1, DNA-damage inducible 1, homolog 1; DDI1, DNA-damage inducible 1, homolog 1 (S. cerevisiae); DNA damage inducible 1 homolog 1; DNA-damage inducible 1 homolog 1; DNA-damage inducible 1 homolog 1 (S. cerevisiae); DNA-damage inducible protein 1; protein DDI1 homolog 1; RGD1559430 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 414301, 71829 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.16 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8WTU0, Q9DAF3 | |
| DDI1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 97-396 of human DDI1 (NP_0010017111). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.