missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDI2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 391.65 - € 590.10
Specifications
| Antigen | DDI2 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18137277
|
Novus Biologicals
NBP2-47492 |
0.1 mL |
€ 624.00 € 590.10 / 0.10mL Save € 33.90 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629095
|
Novus Biologicals
NBP2-47492-25ul |
25 μL |
€ 415.00 € 391.65 / 25µL Save € 23.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DDI2 Polyclonal specifically detects DDI2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| DDI2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DDI1, DNA-damage inducible 1, homolog 2, DDI1, DNA-damage inducible 1, homolog 2 (S. cerevisiae), DNA-damage inducible 1 homolog 2 (S. cerevisiae), DNA-damage inducible protein 2 (DDI2), MGC14844, protein DDI1 homolog 2, RP4-680D5.5 | |
| DDI2 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 84301 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VDADFELHNFRALCELESGIPAAESQIVYAERPLTDNHRSLASYGL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title