missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDI2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560247
This item is not returnable.
View return policy
Description
Immunogen sequence: EGELPECARL AYGAGREDVR PEEIADQELA EALQKSAEDA ERQKP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 84%.
Aspartic protease that mediates the cleavage of NFE2L1/NRF1 at 'Leu-104', thereby promoting release of NFE2L1/NRF1 from the endoplasmic reticulum membrane. Ubiquitination of NFE2L1/NRF1 is a prerequisite for cleavage, suggesting that DDI2 specifically recognizes and binds ubiquitinated NFE2L1/NRF1.
Specifications
| DDI2 | |
| Polyclonal | |
| Unconjugated | |
| DDI2 | |
| 1110056G13Rik; 1700027M01Rik; 9130022E05Rik; AI604911; AU040698; DDI1, DNA-damage inducible 1, homolog 2; Ddi2; DNA damage inducible 1 homolog 2; DNA-damage inducible 1 homolog 2; DNA-damage inducible protein 2; DNA-damage inducible protein 2 (DDI2); protein DDI1 homolog 2; RP4-680D5.5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84301 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5TDH0 | |
| DDI2 | |
| Recombinant protein corresponding to Human DDI2. Recombinant protein control fragment (Product #RP-99835). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction