Learn More
DDIT3 Monoclonal Antibody (OTI2D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA802218S
Encompass_Preferred
 View more versions of this product
Product Code. 17500763
  • € 242.00 / 30µL
Description
DDIT3 Monoclonal antibody specifically detects DDIT3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
DDIT3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q12981
Mouse
Affinity chromatography
RUO
662
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Immunohistochemistry (Paraffin), Western Blot
OTI2D3
Unconjugated
BNIP1
C/EBP homologous protein; C/EBP homoologous protein 10; C/EBP zeta; C/EBP-homologous protein; C/EBP-homologous protein 10; CCAAT/enhancer-binding protein homologous protein; CEBPZ; CHOP; CHOP10; CHOP-10; Ddit3; DDIT-3; ddit3.L; DNA damage inducible transcript 3; DNA damage inducible transcript 3 L homeolog; DNA damage-inducible transcript 3 protein; DNA-damage inducible transcript 3; DNA-damage-inducible transcript 3; GADD153; growth arrest and DNA damage-inducible; growth arrest and DNA damage-inducible protein GADD153; Growth arrest and DNA-damage-inducible protein GADD153; MGC4154; transcription factor GADD153; Xchop; XELAEV_18013440mg
Human recombinant protein fragment corresponding to amino acids 1-199 of human BNIP1 produced in E.coli.
30 μL