missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDIT4 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-22966AF700
This item is not returnable.
View return policy
Description
DDIT4 Polyclonal antibody specifically detects DDIT4 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications
| DDIT4 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Dig2, DNA-damage-inducible transcript 4, FLJ20500, HIF-1 responsive RTP801 (RTP801), Protein regulated in development and DNA damage response 1, REDD-1DNA damage-inducible transcript 4 protein, REDD1HIF-1 responsive protein RTP801, RTP801RP11-442H21.1 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human protein regulated in development and DNA damage response 1 (DNA-damage-inducible transcript 4) using the numbering given in entry NP_061931.1 (GeneID 54541). | |
| 0.1 mL | |
| Cancer, mTOR Pathway | |
| 54541 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction