missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDO Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545190
This item is not returnable.
View return policy
Description
Peptide sequence: TQKQWFRETF NHLFAIANSA EAGDAGVHLV SGWQIFQSTP TEEVPFWADV Sequence homology: Cow: 86%; Dog: 93%; Horse: 93%; Human: 100%; Pig: 93%; Rabbit: 86%.
DDO is a peroxisomal flavoprotein that catalyzes the oxidative deamination of D-aspartate and N-methyl D-aspartate. Flavin adenine dinucleotide or 6-hydroxyflavin adenine dinucleotide can serve as the cofactor in this reaction.
Specifications
| DDO | |
| Polyclonal | |
| Unconjugated | |
| DDO | |
| 5330420D20Rik; 5730402C02Rik; AI467244; aspartic oxidase; DASOX; D-aspartate oxidase; D-aspartate oxidase, DDO; Ddo; DDO protein; DDO-1; DDO-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8528 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99489 | |
| DDO | |
| Synthetic peptide directed towards the N-terminal of human DDO (aa 63-112). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction