missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCD2 Mouse anti-Human, Clone: OTI3B7, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810066
This item is not returnable.
View return policy
Description
ABCD2 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. The function of this peroxisomal membrane protein is unknown; however this protein is speculated to function as a dimerization partner of ABCD1 and/or other peroxisomal ABC transporters.Specifications
| ABCD2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UBJ2 | |
| ABCD2 | |
| Human recombit protein fragment corresponding to amino acids 418-506 of human ABCD2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3B7 | |
| Unconjugated | |
| ABCD2 | |
| ABC39, ALDL1, ALDR, ALDRP, hALDR | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 225 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction