missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDR1 Rabbit anti-Human, Clone: 17, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 405.00 - € 589.00
Specifications
| Antigen | DDR1 |
|---|---|
| Clone | 17 |
| Dilution | Sandwich ELISA Detection 1:1000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18223877
|
Novus Biologicals
NBP2-89617-50UL |
50 μg |
€ 405.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18228456
|
Novus Biologicals
NBP2-89617-100UL |
100 μg |
€ 589.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DDR1 Monoclonal specifically detects DDR1 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
| DDR1 | |
| Sandwich ELISA Detection 1:1000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 780 | |
| IgG | |
| Protein A purified |
| 17 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CD167 antigen-like family member A, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain receptor 1, HGK2, MCK10, MCK-10, NTRK4, Protein-tyrosine kinase 3A, PTK3A, PTK3A protein tyrosine kinase 3A, receptor, type 4, TRK E | |
| Recombinant Human DDR1 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title