missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDR2 Antibody (SR2306), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21783-100ul
This item is not returnable.
View return policy
Description
DDR2 Monoclonal antibody specifically detects DDR2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| DDR2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR2306 | |
| Western Blot 1:500-1:2000 | |
| CD167 antigen-like family member B, CD167b antigen, Discoidin domain receptor 2, discoidin domain receptor family, member 2, discoidin domain receptor tyrosine kinase 2, discoidin domain-containing receptor 2, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, migration-inducing gene 16 protein, neurotrophic tyrosine kinase receptor related 3, Neurotrophic tyrosine kinase, receptor-related 3, NTRKR3cell migration-inducing protein 20, Receptor protein-tyrosine kinase TKT, TKTMIG20a, TYRO10, Tyrosine-protein kinase TYRO10, tyrosylprotein kinase | |
| A synthesized peptide derived from human DDR2 (Uniprot #: Q16832) | |
| 100 μg | |
| Cardiovascular Biology, CD Markers, Immunology, Neuroscience, Signal Transduction | |
| 4921 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction