missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDT Antibody (OTI1H3), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70562AF405
This item is not returnable.
View return policy
Description
DDT Monoclonal specifically detects DDT in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| DDT | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DDT | |
| Full length recombinant protein of human DDT (NP_001346) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1H3 | |
| Western Blot, Immunohistochemistry | |
| D-dopachrome decarboxylase, D-dopachrome tautomeraseDDCT, EC 4.1.1.84, Phenylpyruvate tautomerase II | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 1652.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction