missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDT Monoclonal Antibody (OTI1H3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526176
This item is not returnable.
View return policy
Description
DDT Monoclonal Antibody for Western Blot, IHC (P)
Tautomerization of D-dopachrome with decarboxylation to give 5,6-dihydroxyindole (DHI).
Specifications
| DDT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P30046 | |
| DDT | |
| Full length human recombinant protein of DDT produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H3 | |
| Unconjugated | |
| DDT | |
| C78655; DDCT; D-dopachrome decarboxylase; D-dopachrome tautomerase; Ddt; dopachrome isomerase; hCG_41098; phenylpyruvate tautomerase II | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1652 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction