missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85294
This item is not returnable.
View return policy
Description
DDX1 Polyclonal specifically detects DDX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| DDX1 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| ATP-dependent RNA helicase DDX1, DBP-RBDEAD box polypeptide 1, DEAD (Asp-Glu-Ala-Asp) box polypeptide 1, DEAD box protein 1, DEAD box protein retinoblastoma, DEAD box-1, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 1, EC 3.6.1, EC 3.6.1.10, EC 3.6.4.13, UKVH5d | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of human DDX1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DDX1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NQALFPACVLKNAELKFNFGEEEFKFPPKDGFVALSKAPDGYIVKSQHSGNAQVTQTKFLPNAPKALIVEPSRELAEQTLNNIKQFKKY | |
| 0.1 mL | |
| Cancer, Neuroscience, Transcription Factors and Regulators, Vision | |
| 1653 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction