missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-1576
This item is not returnable.
View return policy
Description
DDX10 Polyclonal specifically detects DDX10 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications
| DDX10 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DDX10-NUP98 fusion protein type 2, DEAD (Asp-Glu-Ala-Asp) box polypeptide 10, DEAD box protein 10, DEAD box-10, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 10 (RNA helicase), EC 3.6.1, EC 3.6.4.13, HRH-J8, probable ATP-dependent RNA helicase DDX10 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 1662 | |
| Store at 4C. Do not freeze. |
| Immunoprecipitation, Western Blot | |
| 1.0 mg/mL | |
| Immunoprecipitation 1-4 ug/mg of lysate, Western Blot (Negative) | |
| NP_004389.2 | |
| DDX10 | |
| The immunogen recognized by this antibody maps to a region between residues 500 and 550 of human DEAD (Asp-Glu-Ala-Asp) Box Polypeptide 10 using the numbering given in entry NP_004389.2 (GeneID 1662). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction