missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX11 Polyclonal Antibody

DDX11 Polyclonal Antibody
Brand: Invitrogen PA523956
This item is not returnable.
View return policy
Description
DDX11 Polyclonal Antibody for Western Blot, Flow
Specifications
| DDX11 | |
| Polyclonal | |
| Unconjugated | |
| DDX11 | |
| 4732462I11Rik; ATP-dependent DNA helicase DDX11; CHL1; CHL1-like helicase homolog; CHL1-related helicase gene-1; CHL1-related protein 1; CHLR1; DDX11; DEAD/H (Asp-Glu-Ala-Asp/His) box helicase 11; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11 (CHL1-like helicase homolog, S. cerevisiae); DEAD/H box protein 11; DEAD/H-box helicase 11; DEAD/H-box protein 11; essa15a; hCHLR1; Keratinocyte growth factor-regulated gene 2 protein; KRG2; KRG-2; probable ATP-dependent DNA helicase DDX11; probable ATP-dependent RNA helicase DDX11; WABS | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 1663, 320209 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6AXC6, Q96FC9 | |
| DDX11 | |
| KLH conjugated synthetic peptide between 656-683 amino acids from the central region of human DDX11. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction