missing translation for 'onlineSavingsMsg'
Learn More

DDX18 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen DDX18
Concentration 400 μg/mL
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16826175
View Documents
Proteintech
28502-1-AP-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16816175
View Documents
Proteintech
28502-1-AP-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The DDX18 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DDX18. This antibody recognizes human, mouse antigen. The DDX18 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

DDX18
Western Blot, ELISA
Unconjugated
Human, Mouse
IgG
400 μg/mL
Polyclonal
Rabbit
8886
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.