missing translation for 'onlineSavingsMsg'
Learn More

DDX19A Monoclonal Antibody (OTI3C12), TrueMAB™, OriGene

Product Code. p-200015607
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17542674 30 μL 30µL
17522674 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17542674 Supplier Origene Technologies Supplier No. TA809971S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DDX19A Monoclonal antibody specifically detects DDX19A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen DDX19A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TEKT2
Gene Accession No. Q9UIF3
Gene Alias ATP-dependent RNA helicase DDX19A; DBP5; Ddx19; DDX19A; DDX19-DDX19L; DDX19L; DDX19-like protein; DEAD (Asp-Glu-Ala-As) box polypeptide 19A; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19a; DEAD box protein 19A; DEAD box RNA helicase DEAD5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19; DEAD-box helicase 19A; Eif4a-rs1; eukaryotic translation initiation factor 4A related sequence 1; eukaryotic translation initiation factor 4A-related sequence 1; mDEAD5; RNA helicase; ZD10B; zinc responsive protein ZD10B
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-244 of human TEKT2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27285
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.