Learn More
DDX19B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand:  Novus Biologicals NB100-760
Product Code. 18708963
  • ONLINE EXCLUSIVE PRICE
    € 601.00
    € 568.05 / 0.10mL
    Save € 32.95
    5% Off
Description
DDX19B Polyclonal specifically detects DDX19B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunohistochemistry
DDX19B
0.1 mL
Store at 4C. Do not freeze.
Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 2- 5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100-1:500
The immunogen recognized by this antibody maps to a region between residue 450 and the C-terminus (residue 479) of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 19B using the numbering given in entry NP_009173.1 (GeneID 11269).