Learn More
DDX19B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB100-760
This item is not returnable.
View return policy
Description
DDX19B Polyclonal specifically detects DDX19B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Western Blot, Immunohistochemistry | |
| DDX19B | |
| 0.1 mL | |
| Store at 4C. Do not freeze. |
| Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 2- 5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100-1:500 | |
| The immunogen recognized by this antibody maps to a region between residue 450 and the C-terminus (residue 479) of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 19B using the numbering given in entry NP_009173.1 (GeneID 11269). | |
| RUO | |
| IgG |