missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX24 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35561-100ul
This item is not returnable.
View return policy
Description
DDX24 Polyclonal antibody specifically detects DDX24 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| DDX24 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| ATP-dependent RNA helicase DDX24, DEAD (Asp-Glu-Ala-Asp) box polypeptide 24, DEAD box protein 24, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24, EC 3.6.1, EC 3.6.4.13, S. cerevisiae CHL1-like helicase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 510-859 of human DDX24 (NP_065147.1).,, Sequence:, LTLVHQAPARILHKKHTKKMDKTAKLDLLMQKIGMRGKPKVIDLTRNEATVETLTETKIHCETDEKDFYLYYFLMQYPGRSLVFANSISCIKRLSGLLKVLDIMPLTLHACMHQKQRLRNLEQFARLEDCVLLATDVAARGLDIPKVQHVIHYQVPRTSEIYVHRSGRTARATNEGLSLMLIGPEDVINFKKIYKTLKKDEDIPLFPVQTKYMDVVKERIRLARQIEKSEYRNFQACLHNSWIEQAAAALEIELEEDMYKGGKADQQEERRRQKQMKVLKKELRHLLSQPLFTESQKTKYPTQSGKPPLLVSAPSKSESALSCLSKQKKKKTKKPKEPQPEQPQPSTSAN | |
| 100 μL | |
| Cell Cycle and Replication | |
| 57062 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction