missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92748-0.02ml
This item is not returnable.
View return policy
Description
DDX3 Polyclonal antibody specifically detects DDX3 in Human, Rat samples. It is validated for Western Blot
Specifications
| DDX3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DBXCAP-Rf, DDX14, DDX3DEAD/H box-3, DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked, DEAD box protein 3, X-chromosomal, DEAD box, X isoform, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, EC 3.6.1, EC 3.6.4.13, helicase like protein 2, Helicase-like protein 2, HLP2ATP-dependent RNA helicase DDX3X | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human DDX3Y (NP_001116137.1). MSHVVVKNDPELDQQLANLDLNSEKQSGGASTASKGRYIPPHLRNREASKGFHDKDSSGWSCSKDKDAYSSFGSRDSRGKPGYFSERGSGSRGRFDDRGRSDYDGIGNRERPGFGRFERSGHSRWCDKSVEDDWSKPLPPSERLEQELFSGGNTGINFEKYDDIPVEATGSNCPPHIENFSDIDMGEIIMGNIELTRYTRPTPVQKHAIP | |
| 0.02 mL | |
| Cancer, Tumor Suppressors | |
| 1654 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction