Learn More
Memmert™ Fresh-Air Filter for IF750
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF750 Incubator
Unit Size:
Each
Description
- Filtration efficiency 80%
- Mounted at the bottom of the appliance
Specifications
Specifications
| For Use With (Equipment) | IF750 Incubator |
| Type | Fresh Air Filter |