Learn More
Memmert™ Fresh-Air Filter for IF750
Product Code. 17527245 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF750 Incubator
Unit Size:
Each
Product Code. 17527245
Brand: Memmert™ IF750R8
Only null left
Add to basket
Add to basket
Fresh-air filter designed to be mounted on the bottom of the unit. Castor frame or subframe not included.
  • Filtration efficiency 80%
  • Mounted at the bottom of the appliance
Specifications
For Use With (Equipment) IF750 Incubator
Type Fresh Air Filter