missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX39B Mouse anti-Canine, Human, Rat, Clone: OTI2E4, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501235
This item is not returnable.
View return policy
Description
This gene encodes a member of the DEAD box family of RNA-dependent ATPases that mediate ATP hydrolysis during pre-mRNA splicing. The encoded protein is an essential splicing factor required for association of U2 small nuclear ribonucleoprotein with pre-mRNA, and also plays an important role in mRNA export from the nucleus to the cytoplasm. A cluster of genes, BAT1-BAT5, is localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. Mutations in this gene may be associated with rheumatoid arthritis. Alternatively spliced transcript variants encoding the same protein have been described.Specifications
| DDX39B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13838, Q5WR10, Q63413 | |
| DDX39B | |
| Full length human recombit protein of human BAT1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2E4 | |
| Unconjugated | |
| DDX39B | |
| BAT1, D6S81E, UAP56 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 114612, 474839, 7919 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction