Learn More
DDX4 VASA Mouse anti-Human, CoraLite 594, Clone: 1F12C3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-67147-100UL
Product Code. 17472683
  • € 450.00 / 100µL
Description
The DDX4, VASA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DDX4, VASA. This antibody recognizes human, mouse antigen. The DDX4, VASA antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
DDX4 VASA
Monoclonal
CoraLite 594
100 μL
54514
IgG1
Immunofluorescence
1F12C3