Learn More
DDX4 VASA Mouse anti-Human, CoraLite 594, Clone: 1F12C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-67147-100UL
Description
The DDX4, VASA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DDX4, VASA. This antibody recognizes human, mouse antigen. The DDX4, VASA antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| DDX4 VASA | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 54514 | |
| IgG1 |
| Immunofluorescence | |
| 1F12C3 | |
| Mouse | |
| Primary | |
| Human, Mouse |