missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX41 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99114CL1
This item is not returnable.
View return policy
Description
DDX41 Polyclonal antibody specifically detects DDX41 in Human samples. It is validated for Western Blot
Specifications
| DDX41 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| ABSDEAD-box protein abstrakt, DEAD (Asp-Glu-Ala-Asp) box polypeptide 41, DEAD box protein 41, DEAD box protein abstrakt homolog, EC 3.6.1, EC 3.6.4.13,2900024F02Rik, MGC8828, probable ATP-dependent RNA helicase DDX41, putative RNA helicase | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human DDX41. | |
| 0.1 mL | |
| Apoptosis | |
| 51428 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction