missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX41 Antibody (PCRP-DDX41-1B4), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08520IR
This item is not returnable.
View return policy
Description
DDX41 Monoclonal antibody specifically detects DDX41 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| DDX41 | |
| Monoclonal | |
| DyLight 755 | |
| ABSDEAD-box protein abstrakt, DEAD (Asp-Glu-Ala-Asp) box polypeptide 41, DEAD box protein 41, DEAD box protein abstrakt homolog, EC 3.6.1, EC 3.6.4.13,2900024F02Rik, MGC8828, probable ATP-dependent RNA helicase DDX41, putative RNA helicase | |
| Recombinant full-length human DDX41 protein (Uniprot: Q9UJV9) | |
| 100 μg | |
| Apoptosis | |
| 51428 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-DDX41-1B4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction