Learn More
Goat anti-Mouse IgG (H+L) Secondary Antibody, PerCP, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP1-72739PCP
Product Code. 30232506
  • ONLINE EXCLUSIVE PRICE
    € 366.00
    € 345.45 / 0.50mL
    Save € 20.55
    5% Off
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot, ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
PBS
Mouse IgG whole molecule
0.5 mL
Adaptive Immunity, Immunology
Mouse
IgG
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot, ChIP Assay
PerCP
Goat
Multi-step
RUO
Secondary
Store at 4°C in the dark.