missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX50 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60676
This item is not returnable.
View return policy
Description
DDX50 Polyclonal specifically detects DDX50 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| DDX50 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| ATP-dependent RNA helicase DDX50, DEAD (Asp-Glu-Ala-Asp) box polypeptide 50, DEAD box protein 50, EC 3.6.1, EC 3.6.4.13, GU2, GUB, gu-beta, MGC3199, Nucleolar protein Gu2, RH-II/GuB, RNA helicase II/Gu beta | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79009 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation | |
| NP_076950.1 | |
| DDX50 | |
| The immunogen recognized by this antibody maps to a region between residue 687 and 737 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 50 using the numbering given in entry NP_076950.1 (GeneID 79009). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction