missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX51 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 463.00
Specifications
| Antigen | DDX51 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18670091
|
Novus Biologicals
NBP2-92254-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649790
|
Novus Biologicals
NBP2-92254-0.1ml |
0.1 mL |
€ 463.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DDX51 Polyclonal antibody specifically detects DDX51 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| DDX51 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| ATP-dependent RNA helicase DDX51, DEAD (Asp-Glu-Ala-Asp) box polypeptide 51, DEAD box protein 51, DKFZp686N2081, EC 3.6.1, EC 3.6.4.13, MGC42193 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 367-666 of human DDX51 (NP_778236.2). FLIIDEADRMIDSMHQSWLPRVVAAAFQSEDPADPCALLQRRQAQAVTAASTCCPQMPLQKLLFSATLTQNPEKLQQLGLHQPRLFSTGLAHRGLEDTDGDGDSGKYAFPVGLTHHYVPCSLSSKPLVVLHLVLEMGFSRVLCFTNSRENSHRLFLLVQAFGGVDVAEFSSRYGPGQRRMILKQFEQGKIQLLISTDATARGIDVQGVELVVNYDAPQYLRTYVHRVGRTARAGKTGQAFTLLLKVQERRFLRMLTEAGAPELQRHELSSKLLQPLVPRYEEALSQLEESVKEERKQRAA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 317781 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title