missing translation for 'onlineSavingsMsg'
Learn More

DDX52 Polyclonal Antibody
GREENER_CHOICE

DDX52 Polyclonal Antibody

Brand:  Invitrogen PA536507

Product Code. 15513901

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DDX52 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DDX52
Polyclonal
Unconjugated
DDX52
2700029C06Rik; ATP-dependent RNA helicase ROK1-like; ATP-dependent, RNA helicase; DDX52; DEAD (Asp-Glu-Ala-Asp) box polypeptide 52; DEAD box protein 52; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 52; DEAD-box helicase 52; HUSSY19; HUSSY-19; probable ATP-dependent RNA helicase DDX52; Rok1; rROK1L
Rabbit
Antigen affinity chromatography
RUO
11056
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.05% sodium azide; pH 7.2
Q9Y2R4
DDX52
Synthetic peptide corresponding to amino acids 553-598 of human DDX52.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.