missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX54 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60678AF350
This item is not returnable.
View return policy
Description
DDX54 Polyclonal antibody specifically detects DDX54 in Human, Mouse samples. It is validated for Western Blot
Specifications
| DDX54 | |
| Polyclonal | |
| Western Blot | |
| APR-5, ATP-dependent RNA helicase DP97, DEAD (Asp-Glu-Ala-Asp) box polypeptide 54, DEAD box helicase 97 KDa, DEAD box protein 54, DEAD box RNA helicase 97 kDa, DP97ATP-dependent RNA helicase DDX54, EC 3.6.1, EC 3.6.4.13, MGC2835 | |
| The immunogen recognized by this antibody maps to a region between residue 800 and 850 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 54 using the numbering given in entry NP_076977.3 (GeneID 79039). | |
| 0.1 mL | |
| Signal Transduction | |
| 79039 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction