missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX56 Monoclonal Antibody (OTI11H7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803232
This item is not returnable.
View return policy
Description
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene shows ATPase activity in the presence of polynucleotides and associates with nucleoplasmic 65S preribosomal particles. This gene may be involved in ribosome synthesis, most likely during assembly of the large 60S ribosomal subunit.Specifications
| DDX56 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NY93 | |
| DDX56 | |
| Human recombit protein fragment corresponding to amino acids 323-547 of human DDX56 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI11H7 | |
| Unconjugated | |
| DDX56 | |
| DDX21, DDX26, NOH61 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54606 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction