missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX56 Monoclonal Antibody (OTI11H7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803232S
This item is not returnable.
View return policy
Description
DDX56 Monoclonal antibody specifically detects DDX56 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| DDX56 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y572 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11035 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI11H7 | |
| Unconjugated | |
| RIPK3 | |
| 2600001H07Rik; 61-kd nucleolar helicase; ATP-dependent 61 kDa nucleolar RNA helicase; D11Ertd619e; DDX21; DDX26; DDX56; DEAD (Asp-Glu-Ala-Asp) box helicase 56; DEAD (Asp-Glu-Ala-Asp) box polypeptide 56; DEAD box protein 21; DEAD box protein 56; DEAD-box helicase 56; DEAD-box RNA helicase; NOH61; nucleolar helicase of 61 kDa; nucleolar RNA helicase homolog; probable ATP-dependent RNA helicase DDX56; putative nucleolar RNA helicase; RH-II/Gu; RNA helicase II/Gu | |
| Human recombinant protein fragment corresponding to amino acids 233-518 of human RIPK3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction