missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX59 Antibody (OTI2G1), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72229UV
This item is not returnable.
View return policy
Description
DDX59 Monoclonal specifically detects DDX59 in Human samples. It is validated for Immunohistochemistry.
Specifications
| DDX59 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DDX59 | |
| Recombinant protein fragment corresponding to amino acids 1-303 of human DDX59 (NP_001026895) produced in E.coli. | |
| 0.1 mL | |
| 83479.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI2G1 | |
| Immunohistochemistry | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 59, DEAD box protein 59, DKFZP564B1023, EC 3.6.1, EC 3.6.4.13, Zinc finger HIT domain-containing protein 5, ZNHIT5probable ATP-dependent RNA helicase DDX59 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction