missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX6 Monoclonal Antibody (OTI3B7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804349S
This item is not returnable.
View return policy
Description
DDX6 Monoclonal antibody specifically detects DDX6 in Human samples. It is validated for Western BlotSpecifications
| DDX6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q00987 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4193 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI3B7 | |
| Unconjugated | |
| MDM2 | |
| 1110001P04Rik; ATP-dependent RNA helicase p54; DDX6; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 6; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 6; DEAD (Asp-Glu-Ala-Asp) box helicase 6; DEAD (Asp-Glu-Ala-Asp) box polypeptide 6; DEAD box protein 6; DEAD box-6; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6 (RNA helicase, 54kD); DEAD-box helicase 6; DEAD-box protein 6; E230023J21Rik; HLR2; mRCK/P54; Oncogene RCK; oncogene RCK homolog; p54; probable ATP-dependent RNA helicase DDX6; RCK; RGD1564560 | |
| Human recombinant protein fragment corresponding to amino acids 119-438 of human MDM2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction