Learn More
Deacetyl Diltiazem N-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | Deacetyl Diltiazem N-Oxide |
| CAS | 122619-90-3 |
| Molecular Formula | C20H24N2O4S |
| Quantity | 10 mg |
| Synonym | (2S,3S)-5-[2-(Dimethyloxidoamino)ethyl]-2,3-dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one, (2S-cis)-5-[2-(Dimethylamino)ethyl]-2,3-dihydro- 3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one N-Oxide |
| InChI Formula | InChI=1S/C20H24N2O4S/c1-22(2,25)13-12-21-16-6-4-5-7-17(16)27-19(18(23)20(21)24)14-8-10-15(26-3)11-9-14/h4-11,18-19,23H,12-13H2,1-3H3/t18-,19+/m1/s1 |
| SMILES | O[C@@H](C(N(CCN(C)(C)=O)C(C=CC=C1)=C1S2)=O)[C@@H]2C3=CC=C(OC)C=C3 |
| IUPAC Name | 2-[(2S,3S)-3-hydroxy-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-5-yl]-N,N-dimethylethanamine oxide |
| Molecular Weight (g/mol) | 388.48 |